Product Details

SNP ID
rs1802619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50317897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAAGGGAGTAGGGTCAGGTCCT[A/C]CCCAGCCCAGCTGTACCCTCCTTCC
Phenotype
MIM: 607071 MIM: 603551 MIM: 607052
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HYAL1 PubMed Links

Gene Details

Gene
HYAL1
Gene Name
hyaluronoglucosaminidase 1
There are no transcripts associated with this gene.

Gene
HYAL2
Gene Name
hyaluronoglucosaminidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003773.4 2022 UTR 3 NP_003764.3
NM_033158.4 2022 UTR 3 NP_149348.2
XM_005265524.2 2022 UTR 3 XP_005265581.1
XM_005265525.2 2022 UTR 3 XP_005265582.1
Gene
TUSC2
Gene Name
tumor suppressor candidate 2
There are no transcripts associated with this gene.

View Full Product Details