Product Details

SNP ID
rs1894324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50243449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTGGAAATGCAGCAACAGCCCC[A/G]AGATGAGCTCACTCTGGGTTTCCCC
Phenotype
MIM: 139360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAI2 PubMed Links

Gene Details

Gene
GNAI2
Gene Name
G protein subunit alpha i2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166425.1 Intron NP_001159897.1
NM_001282617.1 Intron NP_001269546.1
NM_001282618.1 Intron NP_001269547.1
NM_001282619.1 Intron NP_001269548.1
NM_001282620.1 Intron NP_001269549.1
NM_002070.3 Intron NP_002061.1

View Full Product Details