Product Details

SNP ID
rs5266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231925659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGCCTTGTCGCCCTTCTTCTG[A/G]CCGCCGCCCGCAGCCTGCGGCTCGG
Phenotype
MIM: 600296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPPC PubMed Links

Gene Details

Gene
NPPC
Gene Name
natriuretic peptide C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024409.3 285 Silent Mutation GGC,GGT G49G NP_077720.1
XM_011511245.2 285 Silent Mutation GGC,GGT G49G XP_011509547.1

View Full Product Details