Product Details

SNP ID
rs1804246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85659556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAAGTGACTGCCGAGGAAGGG[A/C]GGTGAGGAGGGCGGTGTGGGAGCTG
Phenotype
MIM: 178640
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SFTPB PubMed Links

Gene Details

Gene
SFTPB
Gene Name
surfactant protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000542.3 1428 UTR 3 NP_000533.3
NM_198843.2 1428 Intron NP_942140.2
XM_005264487.2 1428 UTR 3 XP_005264544.1
XM_005264488.3 1428 UTR 3 XP_005264545.2
XM_005264490.4 1428 Intron XP_005264547.2
XM_017004702.1 1428 Intron XP_016860191.1
XM_017004703.1 1428 Intron XP_016860192.1

View Full Product Details