Product Details

SNP ID
rs2044125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63785824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACATGGTTCATAGTACCTTTCCT[G/T]CAGACATGAATACATAGATACTGTC
Phenotype
MIM: 613369
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX42 PubMed Links

Gene Details

Gene
DDX42
Gene Name
DEAD-box helicase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007372.3 Intron NP_031398.2
NM_203499.2 Intron NP_987095.1
XM_006721657.1 Intron XP_006721720.1
XM_011524259.2 Intron XP_011522561.1
XM_017024111.1 Intron XP_016879600.1
XM_017024112.1 Intron XP_016879601.1

View Full Product Details