Product Details

SNP ID
rs1860130
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:109850295 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGGAGATAAGTCTTTTTCCAGGCT[A/G]AAATGAATCTAAATTCTCAGATTCC
Phenotype
MIM: 615004 MIM: 605224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRIT3 PubMed Links

Gene Details

Gene
LRIT3
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198506.4 Intron NP_940908.3
XM_017008167.1 Intron XP_016863656.1
XM_017008168.1 Intron XP_016863657.1
Gene
RRH
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
There are no transcripts associated with this gene.

View Full Product Details