Product Details

SNP ID
rs826534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108725252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGCTGTGCTCCTTCCTAGCTGT[A/G]TATCTTTAAACAAGGCATTTACCTG
Phenotype
MIM: 601181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RANBP2 PubMed Links

Gene Details

Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 Intron NP_006258.3
XM_005264002.2 Intron XP_005264059.1
XM_005264003.2 Intron XP_005264060.1
XM_005264004.2 Intron XP_005264061.1
XM_005264005.4 Intron XP_005264062.1
XM_005264007.2 Intron XP_005264064.1
XM_011511575.2 Intron XP_011509877.1
XM_011511576.2 Intron XP_011509878.1
XM_011511578.2 Intron XP_011509880.1
XM_017004623.1 Intron XP_016860112.1
XM_017004624.1 Intron XP_016860113.1
XM_017004625.1 Intron XP_016860114.1

View Full Product Details