Product Details
- SNP ID
-
rs1864152
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:87608467 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGCCGCTTCCCCCGCTGGTAGCTTC[A/C]TGGCTGTGAGAGGCCATGCGGTGGA
- Phenotype
-
MIM: 605268
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
JPH3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141849784] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JPH3
- Gene Name
- junctophilin 3
View Full Product Details