Product Details

SNP ID
rs1862788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87302820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTTAAGAGGTGGCCAGTTTAGA[A/G]TTTCACAAATAACATTTATTGACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf95 PubMed Links

Gene Details

Gene
C16orf95
Gene Name
chromosome 16 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195124.1 955 UTR 3 NP_001182053.1
NM_001195125.1 955 UTR 3 NP_001182054.1
NM_001256917.1 955 UTR 3 NP_001243846.1

View Full Product Details