Product Details

SNP ID
rs17474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1518096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAATAAACAAGGTAATCTAATCA[A/G]TTCTACAAAGTGTAGTTTGTAAAAC
Phenotype
MIM: 607875 MIM: 600174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5K PubMed Links

Gene Details

Gene
INPP5K
Gene Name
inositol polyphosphate-5-phosphatase K
There are no transcripts associated with this gene.

Gene
PITPNA
Gene Name
phosphatidylinositol transfer protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006224.3 3534 UTR 3 NP_006215.1
XM_011523939.1 3534 UTR 3 XP_011522241.1
XM_011523940.1 3534 UTR 3 XP_011522242.1
Gene
PITPNA-AS1
Gene Name
PITPNA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details