Product Details

SNP ID
rs12430
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:75945874 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGTCAAAGGGTTTTTCTGTGCTAC[A/G]TCTGGGAGTGCTGACCTAAGTGACA
Phenotype
MIM: 609751 MIM: 616807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOX1 PubMed Links

Gene Details

Gene
ACOX1
Gene Name
acyl-CoA oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185039.1 3218 UTR 3 NP_001171968.1
NM_004035.6 3218 UTR 3 NP_004026.2
NM_007292.5 3218 UTR 3 NP_009223.2
XM_011524868.2 3218 UTR 3 XP_011523170.1
XM_011524869.2 3218 UTR 3 XP_011523171.1
Gene
FBF1
Gene Name
Fas binding factor 1
There are no transcripts associated with this gene.

View Full Product Details