Product Details

SNP ID
rs2063792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78384689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTATGGAGAGCGCAACTTCTG[C/T]CCTGTTCAGTGAGGACAGCCCTCCA
Phenotype
MIM: 614942
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGS1 PubMed Links

Gene Details

Gene
PGS1
Gene Name
phosphatidylglycerophosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024419.4 Intron NP_077733.3
XM_011525487.2 Intron XP_011523789.1
XM_011525488.2 Intron XP_011523790.1
XM_011525489.2 Intron XP_011523791.1
XM_017025357.1 Intron XP_016880846.1
XM_017025358.1 Intron XP_016880847.1
XM_017025359.1 Intron XP_016880848.1
XM_017025360.1 Intron XP_016880849.1
XM_017025361.1 Intron XP_016880850.1
XM_017025362.1 Intron XP_016880851.1
XM_017025363.1 Intron XP_016880852.1
XM_017025364.1 Intron XP_016880853.1
XM_017025365.1 Intron XP_016880854.1
XM_017025366.1 Intron XP_016880855.1

View Full Product Details