Product Details

SNP ID
rs14526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78853099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAACTCTTGTGTGTTCCCAGCAC[C/G]GGAAGGTCTCAGACTTCATATTCCA
Phenotype
MIM: 188825
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TIMP2 PubMed Links

Gene Details

Gene
TIMP2
Gene Name
TIMP metallopeptidase inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003255.4 3533 UTR 3 NP_003246.1

View Full Product Details