Product Details

SNP ID
rs2859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:107376998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTTGTGTGTGGGTTTTTTTTT[G/T]TTACATTTTCTTTTACGTTTATATA
Phenotype
MIM: 601535
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFNA5 PubMed Links

Gene Details

Gene
EFNA5
Gene Name
ephrin A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001962.2 6614 UTR 3 NP_001953.1
XM_006714565.2 6614 UTR 3 XP_006714628.1
XM_011543250.2 6614 UTR 3 XP_011541552.1
XM_011543251.2 6614 UTR 3 XP_011541553.1
XM_017009205.1 6614 UTR 3 XP_016864694.1

View Full Product Details