Product Details

SNP ID
rs1052589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12730671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAGTCAAGTTCTTTACTTCCCA[A/G]AAGTGATGGCTAAGGGGAGGGAGGA
Phenotype
MIM: 601913 MIM: 603002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASNA1 PubMed Links

Gene Details

Gene
ASNA1
Gene Name
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
There are no transcripts associated with this gene.

Gene
C19orf43
Gene Name
chromosome 19 open reading frame 43
There are no transcripts associated with this gene.

Gene
TNPO2
Gene Name
transportin 2
There are no transcripts associated with this gene.

View Full Product Details