Product Details

SNP ID
rs1045980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10730658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGAGTTAGTATGTTCAACAGA[C/T]CCCATTAGCAGAAGTCATGTTCCAG
Phenotype
MIM: 615318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM14C PubMed Links

Gene Details

Gene
TMEM14C
Gene Name
transmembrane protein 14C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165258.1 702 Missense Mutation CCC,TCC P111S NP_001158730.1
NM_016462.3 702 Missense Mutation CCC,TCC P111S NP_057546.1

View Full Product Details