Product Details

SNP ID
rs13960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3762700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTGCAAGCTGCCCGCAATACGT[C/T]ATGGCGACCAAACGCCTTTTCGGGG
Phenotype
MIM: 604262 MIM: 611858 MIM: 610362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
There are no transcripts associated with this gene.

Gene
MRPL54
Gene Name
mitochondrial ribosomal protein L54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172251.2 34 UTR 5 NP_758455.1
Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
There are no transcripts associated with this gene.

View Full Product Details