Product Details

SNP ID
rs13849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11183663 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCACTGATTTTTTTTTAATCAC[G/T]CTGAGATCATGCATTCTTGTTTTTT
Phenotype
MIM: 602265
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NEDD9 PubMed Links

Gene Details

Gene
NEDD9
Gene Name
neural precursor cell expressed, developmentally down-regulated 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142393.1 4171 UTR 3 NP_001135865.1
NM_001271033.1 4171 UTR 3 NP_001257962.1
NM_006403.3 4171 UTR 3 NP_006394.1
NM_182966.3 4171 Intron NP_892011.2

View Full Product Details