Product Details

SNP ID
rs10875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6210729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAATCACAAAGTGCCAGGCCC[A/C]TTTCCTATGGAAGTGTCCCCAGAGC
Phenotype
MIM: 159556
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MLLT1 PubMed Links

Gene Details

Gene
MLLT1
Gene Name
MLLT1, super elongation complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005934.3 4131 UTR 3 NP_005925.2
XM_011528021.1 4131 UTR 3 XP_011526323.1
XM_011528022.1 4131 UTR 3 XP_011526324.1
XM_011528023.1 4131 UTR 3 XP_011526325.1
XM_017026819.1 4131 UTR 3 XP_016882308.1

View Full Product Details