Product Details

SNP ID
rs285236
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16188363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGTTTACTGGGGTGGGAAGGAT[C/T]GCAGGTAAAGCAGGTTTTCTGGAGT
Phenotype
MIM: 603535 MIM: 614554
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AP1M1 PubMed Links

Gene Details

Gene
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
There are no transcripts associated with this gene.

Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014077.3 Intron NP_054796.1
XM_011527898.1 Intron XP_011526200.1

View Full Product Details