Product Details

SNP ID
rs2027132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:8074246 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTAATTTCCTAACTTCTTTTAC[A/G]TGAGAAAAAAACAACCCACCCTGAT
Phenotype
MIM: 607289 MIM: 609206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S5 PubMed Links

Gene Details

Gene
BLOC1S5
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 5
There are no transcripts associated with this gene.

Gene
BLOC1S5-TXNDC5
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
EEF1E1
Gene Name
eukaryotic translation elongation factor 1 epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135650.1 Intron NP_001129122.1
NM_004280.4 Intron NP_004271.1
Gene
EEF1E1-BLOC1S5
Gene Name
EEF1E1-BLOC1S5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details