Product Details

SNP ID
rs1578989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44444901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTGCTCTACTATTAAAACATT[C/T]TATCCTTTCATTCATAATCTGACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF445 PubMed Links

Gene Details

Gene
ZNF445
Gene Name
zinc finger protein 445
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181489.5 4212 UTR 3 NP_852466.1
XM_005265102.2 4212 UTR 3 XP_005265159.1
XM_005265103.3 4212 UTR 3 XP_005265160.1
XM_011533674.2 4212 UTR 3 XP_011531976.1
XM_011533675.2 4212 UTR 3 XP_011531977.1
XM_017006345.1 4212 UTR 3 XP_016861834.1
XM_017006346.1 4212 UTR 3 XP_016861835.1

View Full Product Details