Product Details
- SNP ID
-
hCV11969451
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52546076 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTATAGCACCTCTAGGCACTGG[A/G]AAGGAGCCTGGAGGAGAGCTCTGGC
- Phenotype
-
MIM: 606083
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PBRM1
PubMed Links
Gene Details
- Gene
- PBRM1
- Gene Name
- polybromo 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018313.4 |
6850 |
UTR 3 |
|
|
NP_060783.3 |
- Gene
- SMIM4
- Gene Name
- small integral membrane protein 4
There are no transcripts associated with this gene.
View Full Product Details