Product Details

SNP ID
rs13673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58565486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAGAATTGCATCGTAACAGTGT[A/G]GTCACACTGGTAAGAAATGCAGATT
Phenotype
MIM: 608295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM107A PubMed Links

Gene Details

Gene
FAM107A
Gene Name
family with sequence similarity 107 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076778.2 2246 UTR 3 NP_001070246.1
NM_001282713.1 2246 UTR 3 NP_001269642.1
NM_001282714.1 2246 UTR 3 NP_001269643.1
NM_007177.3 2246 UTR 3 NP_009108.1

View Full Product Details