Product Details
- SNP ID
-
rs200376
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18177080 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGGCTTGGCTCAGGGCTGGTGGG[C/T]GGGACCGTTGAGATAACGGGGAGGA
- Phenotype
-
MIM: 604664
MIM: 603157
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFI30
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140154009] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFI30
- Gene Name
- IFI30, lysosomal thiol reductase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006332.4 |
|
Intron |
|
|
NP_006323.2 |
- Gene
- PIK3R2
- Gene Name
- phosphoinositide-3-kinase regulatory subunit 2
There are no transcripts associated with this gene.
View Full Product Details