Product Details

SNP ID
rs14567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57090026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATGCAGAAATTCTAAGGTACAA[A/C]AACATTTTGTAAATGTCAGCTGTGA
Phenotype
MIM: 606807
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL17RD PubMed Links

Gene Details

Gene
IL17RD
Gene Name
interleukin 17 receptor D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318864.1 8709 UTR 3 NP_001305793.1
NM_017563.4 8709 UTR 3 NP_060033.3
XM_005265238.4 8709 UTR 3 XP_005265295.1
XM_006713209.3 8709 UTR 3 XP_006713272.1
XM_011533849.2 8709 UTR 3 XP_011532151.2

View Full Product Details