Product Details
- SNP ID
-
rs2035655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:57091174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATATTGTGAGTCATGAAATAAAGG[G/T]GATATCTATAGAGATGTGTAAAAAT
- Phenotype
-
MIM: 606807
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL17RD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77649112] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL17RD
- Gene Name
- interleukin 17 receptor D
View Full Product Details