Product Details

SNP ID
rs1857514
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:110484413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATGAATACATGCTTAACACAC[A/G]TTAATTATTTATTTCTCCACCAATC
Phenotype
MIM: 118943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYMP PubMed Links

Gene Details

Gene
CYMP
Gene Name
chymosin pseudogene
There are no transcripts associated with this gene.

Gene
LOC440602
Gene Name
uncharacterized LOC440602
There are no transcripts associated with this gene.

View Full Product Details