Product Details

SNP ID
rs1834
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:110039158 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTTTCTAGGCTCAGGTGTAACTGGT[G/T]TCTTGCCCTGCAGTGACAGACAAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STRIP1 PubMed Links

Gene Details

Gene
STRIP1
Gene Name
striatin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270768.1 Intron NP_001257697.1
NM_033088.3 Intron NP_149079.2
XM_006710995.2 Intron XP_006711058.1
XM_017002629.1 Intron XP_016858118.1
XM_017002630.1 Intron XP_016858119.1

View Full Product Details