Product Details
- SNP ID
-
rs1048032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:196554275 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGATCAGCTACTAATATATTTTGG[G/T]GTCCCTTTATTTTTGTACCCAAGAG
- Phenotype
-
MIM: 615110
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR53
PubMed Links
Gene Details
- Gene
- WDR53
- Gene Name
- WD repeat domain 53
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182627.1 |
1484 |
Missense Mutation |
CAC,CCC |
H338P |
NP_872433.1 |
View Full Product Details