Product Details

SNP ID
rs5879
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:173903934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCTGCCCATGAAGATAATAGT[A/G]TTCAGAGGAACTTCTCTTATAAAAA
Phenotype
MIM: 107300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 1469 Silent Mutation AAC,AAT N450N NP_000479.1
XM_005245198.3 1469 Silent Mutation AAC,AAT N402N XP_005245255.1

View Full Product Details