Product Details
- SNP ID
-
rs1938564
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:98007323 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACAACTTGGAAGCAGGCCTAACTG[C/T]TCAATTTTATCCAAATAACTAAAGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR137HG
PubMed Links
Gene Details
- Gene
- MIR137HG
- Gene Name
- MIR137 host gene
There are no transcripts associated with this gene.
View Full Product Details