Product Details

SNP ID
rs1134866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48294550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCCAGCAGAAATGGCACTGTAA[C/G]CCAGGCTTCCCTGGTCCTAGGAGAA
Phenotype
MIM: 608294
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCF1P2 PubMed Links

Gene Details

Gene
FCF1P2
Gene Name
FCF1 pseudogene 2
There are no transcripts associated with this gene.

Gene
NME6
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308426.1 988 UTR 3 NP_001295355.1
NM_001308427.1 988 UTR 3 NP_001295356.1
NM_001308428.1 988 UTR 3 NP_001295357.1
NM_001308430.1 988 UTR 3 NP_001295359.1
NM_001308431.1 988 UTR 3 NP_001295360.1
NM_001308433.1 988 UTR 3 NP_001295362.1
NM_001308434.1 988 UTR 3 NP_001295363.1
NM_001308435.1 988 UTR 3 NP_001295364.1
NM_005793.4 988 UTR 3 NP_005784.1
XM_017005511.1 988 Intron XP_016861000.1
XM_017005512.1 988 Intron XP_016861001.1
XM_017005513.1 988 UTR 3 XP_016861002.1
XM_017005514.1 988 UTR 3 XP_016861003.1
XM_017005515.1 988 UTR 3 XP_016861004.1
XM_017005516.1 988 Intron XP_016861005.1
XM_017005517.1 988 UTR 3 XP_016861006.1
XM_017005518.1 988 UTR 3 XP_016861007.1

View Full Product Details