Product Details

SNP ID
rs1721499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105539309 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCTATCCCATCACCAAGTCATC[G/T]GAGTTCAGCCTCAACATCTCTGGTA
Phenotype
MIM: 610089
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RINT1 PubMed Links

Gene Details

Gene
RINT1
Gene Name
RAD50 interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021930.4 Intron NP_068749.3
XM_005250524.3 Intron XP_005250581.1
XM_011516458.2 Intron XP_011514760.1
XM_017012496.1 Intron XP_016867985.1
XM_017012497.1 Intron XP_016867986.1
XM_017012498.1 Intron XP_016867987.1

View Full Product Details