Product Details

SNP ID
rs14522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135674922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGATGTTATATGATGGATAAG[G/T]GAAAAGTTTTTAAAGGAGATTTTAT
Phenotype
MIM: 604309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf73 PubMed Links

Gene Details

Gene
C7orf73
Gene Name
chromosome 7 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130929.1 1008 UTR 3 NP_001124401.1
Gene
SLC13A4
Gene Name
solute carrier family 13 member 4
There are no transcripts associated with this gene.

View Full Product Details