Product Details

SNP ID
rs2001583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129623556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCAAAGGTATCTTTCTTAAAAA[A/T]TTTATATGGTAGAATCCTATTTAGA
Phenotype
MIM: 600879
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NRF1 PubMed Links

Gene Details

Gene
NRF1
Gene Name
nuclear respiratory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040110.1 Intron NP_001035199.1
NM_001293163.1 Intron NP_001280092.1
NM_001293164.1 Intron NP_001280093.1
NM_005011.4 Intron NP_005002.3

View Full Product Details