Product Details
- SNP ID
-
hCV11996705
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:74028188 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGGCTGGGGCATTTCTCCCCGAG[A/G]TGGCGGGTCTGACGGCGGCGGCCCC
- Phenotype
-
MIM: 130160
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELN
PubMed Links
Gene Details
- Gene
- ELN
- Gene Name
- elastin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000501.3 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_000492.2 |
NM_001081752.2 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001075221.1 |
NM_001081753.2 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001075222.1 |
NM_001081754.2 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001075223.1 |
NM_001081755.2 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001075224.1 |
NM_001278912.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265841.1 |
NM_001278913.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265842.1 |
NM_001278914.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265843.1 |
NM_001278915.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265844.1 |
NM_001278916.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265845.1 |
NM_001278917.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265846.1 |
NM_001278918.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265847.1 |
NM_001278939.1 |
400 |
Missense Mutation |
ATG,GTG |
M1V |
NP_001265868.1 |
View Full Product Details