Product Details

SNP ID
rs1805116
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:91683539 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAATTCTAGTGTGGTACAGAAGCC[C/T]AGGGTCATGTTTATACTAGCCCTGG
Phenotype
MIM: 600742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGFBR3 PubMed Links

Gene Details

Gene
TGFBR3
Gene Name
transforming growth factor beta receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195683.1 3271 UTR 3 NP_001182612.1
NM_001195684.1 3271 UTR 3 NP_001182613.1
NM_003243.4 3271 UTR 3 NP_003234.2
XM_006710867.2 3271 UTR 3 XP_006710930.1
XM_006710868.2 3271 UTR 3 XP_006710931.1

View Full Product Details