Product Details

SNP ID
rs953
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:231332860 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTACATAGACTGACTTTTATAGTAC[A/G]TGTCATGTCCCAAATTCCCAATCCT
Phenotype
MIM: 615283 MIM: 616086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC8 PubMed Links

Gene Details

Gene
EXOC8
Gene Name
exocyst complex component 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175876.4 5013 UTR 3 NP_787072.2
Gene
SPRTN
Gene Name
SprT-like N-terminal domain
There are no transcripts associated with this gene.

View Full Product Details