Product Details

SNP ID
rs2035081
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:56732095 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGTGGCAAGATAATTTGTATTGCT[A/G]TAACATTGTCACAGTTTTCTAGAGT
Phenotype
MIM: 601234 MIM: 176635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NACA PubMed Links

Gene Details

Gene
NACA
Gene Name
nascent polypeptide-associated complex alpha subunit
There are no transcripts associated with this gene.

Gene
PRIM1
Gene Name
primase (DNA) subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000946.2 Intron NP_000937.1

View Full Product Details