Product Details

SNP ID
rs1901173
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:139314466 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGTGTTTTCAAAACAAGCAATGTA[A/G]CAGAATTTACCTGTACATGATTTAA
Phenotype
MIM: 608000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA15 PubMed Links

Gene Details

Gene
NAA15
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057175.3 Intron NP_476516.1
XM_005263236.2 Intron XP_005263293.1

View Full Product Details