Product Details

SNP ID
rs2039824
Assay Type
Validated
NCBI dbSNP Submissions
55
Location
Chr.1:248349335 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCCCTCTCTGCTGAAGCTCTCTTG[C/T]TCTGACACCTTCAGCAATGAGGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14C36 PubMed Links

Gene Details

Gene
OR14C36
Gene Name
olfactory receptor family 14 subfamily C member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001918.1 561 Silent Mutation TGC,TGT C187C NP_001001918.1

View Full Product Details