Product Details

SNP ID
hCV12016089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19975836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTTTGCTACCCCTAAAATGAT[G/T]GCAGACTTTCTGGTTGAGCGCAAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K15 PubMed Links

Gene Details

Gene
OR4K15
Gene Name
olfactory receptor family 4 subfamily K member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005486.1 318 Missense Mutation ATG,ATT M106I NP_001005486.1

View Full Product Details