Product Details

SNP ID
rs375839440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61249661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGGGAGCTTAACTTGCTTCTT[A/G]CCCTCAGCATCACCATGAACGGAGA
Phenotype
MIM: 169730 MIM: 611115
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGA5 PubMed Links

Gene Details

Gene
PGA5
Gene Name
pepsinogen 5, group I (pepsinogen A)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014224.4 Intron NP_055039.1
Gene
VWCE
Gene Name
von Willebrand factor C and EGF domains
There are no transcripts associated with this gene.

View Full Product Details