Product Details

SNP ID
rs1901514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:102375576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAAGCAGTCAATAAAAACTCAG[A/G]GAGTTTCTAGCAGTTGGAATCAACA
Phenotype
MIM: 613365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO6A1 PubMed Links

Gene Details

Gene
SLCO6A1
Gene Name
solute carrier organic anion transporter family member 6A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289002.1 Intron NP_001275931.1
NM_001289004.1 Intron NP_001275933.1
NM_001308014.1 Intron NP_001294943.1
NM_173488.4 Intron NP_775759.3
XM_005271874.3 Intron XP_005271931.1
XM_011543147.2 Intron XP_011541449.1
XM_011543148.2 Intron XP_011541450.1
XM_011543150.1 Intron XP_011541452.1
XM_011543153.2 Intron XP_011541455.1
XM_011543154.2 Intron XP_011541456.1

View Full Product Details