Product Details

SNP ID
rs1104990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136995886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGCCAGGGCTCCAGCTTGGTC[A/G]GCGCGACTTCCAAGTGCCCAGTGAG
Phenotype
MIM: 616315 MIM: 606533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004669.2 Intron NP_004660.2

View Full Product Details