Product Details

SNP ID
rs1228493
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225486901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGCAGAAGAATGAAGCAGTT[A/T]AAACCAGCAAAGCTGCAAAGTAGGA
Phenotype
MIM: 609061
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 12869 UTR 3 NP_001008493.1
NM_018212.4 12869 UTR 3 NP_060682.2
XM_005273187.2 12869 UTR 3 XP_005273244.1
XM_011544229.2 12869 Intron XP_011542531.2
XM_017001746.1 12869 UTR 3 XP_016857235.1
XM_017001747.1 12869 Intron XP_016857236.1
XM_017001748.1 12869 Intron XP_016857237.1
XM_017001749.1 12869 Intron XP_016857238.1
XM_017001750.1 12869 Intron XP_016857239.1
XM_017001751.1 12869 UTR 3 XP_016857240.1
XM_017001752.1 12869 Intron XP_016857241.1
XM_017001753.1 12869 UTR 3 XP_016857242.1
XM_017001754.1 12869 UTR 3 XP_016857243.1
XM_017001755.1 12869 UTR 3 XP_016857244.1
XM_017001756.1 12869 UTR 3 XP_016857245.1
XM_017001757.1 12869 UTR 3 XP_016857246.1

View Full Product Details