Product Details
- SNP ID
-
rs1533591
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
47
- Location
-
Chr.1:224115976 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTAGAGTTAAAACACATCCCACAA[C/T]TTATGAATAAATGACATCTCTGTCA
- Phenotype
-
MIM: 609100
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114786314] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO28
- Gene Name
- F-box protein 28
View Full Product Details