Product Details

SNP ID
rs2035897
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:224115762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAATTCGCGCTGTACATATAC[A/G]GTGTTTGGAAAATTGGGCATATAAC
Phenotype
MIM: 609100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO28 PubMed Links

Gene Details

Gene
FBXO28
Gene Name
F-box protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136115.2 Intron NP_001129587.1
NM_015176.3 Intron NP_055991.1

View Full Product Details