Product Details

SNP ID
rs1539378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44135358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCAGGAGAGGGACCGAGTGGA[A/G]CAGGAATAGTATGTGGGGCACCAGC
Phenotype
MIM: 611827
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL14 PubMed Links

Gene Details

Gene
MRPL14
Gene Name
mitochondrial ribosomal protein L14
There are no transcripts associated with this gene.

Gene
TMEM63B
Gene Name
transmembrane protein 63B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318792.1 475 Silent Mutation GAA,GAG E90E NP_001305721.1
NM_018426.2 475 Silent Mutation GAA,GAG E90E NP_060896.1
XM_005249211.3 475 Silent Mutation GAA,GAG E99E XP_005249268.1
XM_005249213.4 475 Silent Mutation GAA,GAG E90E XP_005249270.1
XM_005249214.4 475 Intron XP_005249271.1
XM_005249215.4 475 Intron XP_005249272.1
XM_005249217.1 475 Silent Mutation GAA,GAG E90E XP_005249274.1
XM_006715135.1 475 Silent Mutation GAA,GAG E90E XP_006715198.1
XM_017010999.1 475 Silent Mutation GAA,GAG E99E XP_016866488.1
XM_017011000.1 475 Silent Mutation GAA,GAG E90E XP_016866489.1
XM_017011001.1 475 Intron XP_016866490.1
XM_017011002.1 475 Intron XP_016866491.1

View Full Product Details